Assay Details
Gene Symbol: Cald1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145575.3 | NP_663550.1 | 7 - 8 | 1393 | 82 | |
GenBank mRNA | AK148009.1 | - | 5 - 6 | 765 | 82 | |
BC019435.1 | - | 7 - 8 | 1352 | 5148205 | 82 | |
BQ769983.1 | - | 4 - 5 | 577 | 82 | ||
CX568419.1 | - | 2 - 3 | 89 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
