Assay Details
Gene Symbol: Dennd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172477.4 | NP_766065.1 | 1 - 2 | 262 | 78 | |
GenBank mRNA | AK048041.1 | - | 1 - 2 | 246 | 78 | |
AK081176.1 | - | 1 - 2 | 263 | 78 | ||
AK165202.1 | - | 1 - 2 | 118 | 78 | ||
AK167298.1 | - | 1 - 2 | 116 | 78 | ||
BC056367.1 | - | 1 - 2 | 141 | 78 | ||
BC057310.1 | - | 1 - 2 | 141 | 78 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK081176.1, AK167298.1, NM_172477.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
