Assay Details
Gene Symbol: Dennd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172477.4 | NP_766065.1 | 4 - 5 | 1616 | 106 | |
GenBank mRNA | AK048041.1 | - | 4 - 5 | 1601 | 106 | |
AK081176.1 | - | 4 - 5 | 1617 | 106 | ||
AK135781.1 | - | 2 - 3 | 161 | 106 | ||
AK160784.1 | - | 4 - 5 | 1378 | 106 | ||
AK165202.1 | - | 4 - 5 | 1472 | 106 | ||
AK167298.1 | - | 4 - 5 | 1470 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
