Assay Details
Gene Symbol: Cdc25c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK141737.1 | - | 3 - 4 | 542 | 65 | |
AK168287.1 | - | 3 - 4 | 537 | 65 | ||
BC004669.1 | - | 2 - 3 | 246 | 3498846 | 65 | |
L16926.1 | - | 2 - 3 | 670 | 65 | ||
U15562.1 | - | 3 - 4 | 508 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
