Assay Details
Gene Symbol: Usp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301628.1 | NP_001288557.1 | 14 - 15 | 1974 | 91 | |
NM_027604.4 | NP_081880.2 | 15 - 16 | 2061 | 91 | ||
GenBank mRNA | AF468037.1 | - | 15 - 16 | 1965 | 91 | |
AK014891.1 | - | 9 - 10 | 1272 | 91 | ||
AK122301.1 | - | 14 - 15 | 1889 | 91 | ||
AK133852.1 | - | 14 - 15 | 1883 | 91 | ||
AK145749.1 | - | 15 - 16 | 1965 | 91 | ||
AK161469.1 | - | 15 - 16 | 2004 | 91 | ||
AK161957.1 | - | 14 - 15 | 1922 | 91 | ||
AK163315.1 | - | 14 - 15 | 1903 | 91 | ||
AK168755.1 | - | 14 - 15 | 1909 | 91 | ||
BC003978.1 | - | 2 - 3 | 320 | 91 | ||
BC050042.1 | - | 15 - 16 | 1977 | 30022307 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
