Assay Details
Gene Symbol: E2f3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289920.1 | NP_001276849.1 | 3 - 4 | 578 | 93 | |
NM_010093.3 | NP_034223.1 | 3 - 4 | 1096 | 93 | ||
GenBank mRNA | AF015948.1 | - | 3 - 4 | 398 | 93 | |
AK129048.1 | - | 2 - 3 | 294 | 93 | ||
AK133828.1 | - | 3 - 4 | 1098 | 93 | ||
AK154168.1 | - | 3 - 4 | 538 | 93 | ||
AK165892.1 | - | 3 - 4 | 579 | 93 | ||
BC024125.1 | - | 3 - 4 | 335 | 93 | ||
BC059262.1 | - | 3 - 4 | 495 | 93 | ||
BU705601.1 | - | 3 - 4 | 496 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
