Assay Details
Gene Symbol: E2f3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289920.1 | NP_001276849.1 | 5 - 6 | 859 | 67 | |
NM_010093.3 | NP_034223.1 | 5 - 6 | 1377 | 67 | ||
GenBank mRNA | AF015948.1 | - | 5 - 6 | 679 | 67 | |
AK129048.1 | - | 4 - 5 | 575 | 67 | ||
AK133828.1 | - | 5 - 6 | 1379 | 67 | ||
AK154168.1 | - | 6 - 7 | 864 | 67 | ||
AK165892.1 | - | 5 - 6 | 860 | 67 | ||
BC024125.1 | - | 5 - 6 | 616 | 67 | ||
BC059262.1 | - | 5 - 6 | 776 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
