Assay Details
Gene Symbol: Ankhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175375.3 | NP_780584.2 | 22 - 23 | 4246 | 99 | |
GenBank mRNA | AK017018.1 | - | 12 - 13 | 1585 | 99 | |
AK029322.1 | - | 15 - 16 | 2661 | 99 | ||
AK042754.1 | - | 9 - 10 | 1221 | 99 | ||
AK131166.1 | - | 22 - 23 | 4108 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
