Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100026105.mm.1 |
TC1100002871.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100026105.mm.1 |
NM_001039092.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026105.mm.1 |
NM_001039092.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026105.mm.1 |
NM_153080.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026105.mm.1 |
NM_153080.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026108.mm.1 |
TC1100002871.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100026108.mm.1 |
NM_001039092.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026108.mm.1 |
NM_001039092.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026108.mm.1 |
NM_153080.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026108.mm.1 |
NM_153080.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026104.mm.1 |
TC1100002871.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100026104.mm.1 |
NM_001039092.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026104.mm.1 |
NM_001039092.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026104.mm.1 |
NM_153080.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026104.mm.1 |
NM_153080.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026110.mm.1 |
TC1100002871.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100026110.mm.1 |
NM_153080.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026110.mm.1 |
NM_153080.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026106.mm.1 |
TC1100002871.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100026106.mm.1 |
NM_001039092.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026106.mm.1 |
NM_001039092.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026106.mm.1 |
NM_153080.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026106.mm.1 |
NM_153080.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026107.mm.1 |
TC1100002871.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100026107.mm.1 |
NM_001039092.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026107.mm.1 |
NM_001039092.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026107.mm.1 |
NM_153080.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100026107.mm.1 |
NM_153080.2 |
Refseq |