Assay Details
Gene Symbol: Atp1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290469.1 | NP_001277398.1 | 14 - 15 | 2122 | 68 | |
GenBank mRNA | BC020177.1 | - | 14 - 15 | 2080 | 4506376 | 68 |
BC027000.1 | - | 7 - 8 | 1142 | 68 | ||
BC027114.1 | - | 2 - 3 | 251 | 68 | ||
BC034645.1 | - | 14 - 15 | 2078 | 5357731 | 68 | |
BC037206.1 | - | 14 - 15 | 2077 | 5362423 | 68 | |
BC042894.1 | - | 14 - 15 | 2122 | 5364812 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
