Assay Details
Gene Symbol: Dnm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039520.2 | NP_001034609.1 | 17 - 18 | 2083 | 83 | |
NM_001253893.1 | NP_001240822.1 | 17 - 18 | 2083 | 83 | ||
NM_001253894.1 | NP_001240823.1 | 17 - 18 | 2083 | 83 | ||
NM_007871.2 | NP_031897.2 | 16 - 17 | 2071 | 83 | ||
GenBank mRNA | AK005012.2 | - | 17 - 18 | 2059 | 83 | |
AK157844.1 | - | 17 - 18 | 2052 | 83 | ||
AK170574.1 | - | 17 - 18 | 2059 | 83 | ||
AK171049.1 | - | 17 - 18 | 2037 | 83 | ||
AK171566.1 | - | 17 - 18 | 2048 | 83 | ||
AK172231.1 | - | 17 - 18 | 2064 | 83 | ||
BC010233.1 | - | 8 - 9 | 799 | 3499181 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
