Assay Details
Gene Symbol: Naalad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028279.3 | NP_082555.2 | 6 - 7 | 851 | 67 | |
GenBank mRNA | AK133611.1 | - | 6 - 7 | 961 | 67 | |
AK135653.1 | - | 8 - 9 | 994 | 67 | ||
AK161457.1 | - | 9 - 10 | 1157 | 67 | ||
AY243507.1 | - | 6 - 7 | 851 | 67 | ||
BC108995.1 | - | 6 - 7 | 835 | 40047859 | 67 | |
BC108996.1 | - | 6 - 7 | 835 | 40047860 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
