Assay Details
Gene Symbol: Usf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011680.2 | NP_035810.1 | 7 - 8 | 776 | 74 | |
GenBank mRNA | AB221562.1 | - | 7 - 8 | 735 | 74 | |
AK146871.1 | - | 7 - 8 | 723 | 74 | ||
AK158906.1 | - | 5 - 6 | 552 | 74 | ||
AK211311.1 | - | 2 - 3 | 169 | 74 | ||
BC019729.1 | - | 2 - 3 | 156 | 74 | ||
BC082995.1 | - | 7 - 8 | 776 | 6440967 | 74 | |
U01662.1 | - | 7 - 8 | 792 | 74 | ||
U01663.1 | - | 4 - 5 | 329 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of transcription from RNA polymerase II promoter by glucose
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription by RNA polymerase II
transcription by RNA polymerase II
lactation
positive regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
lipid homeostasis
RNA polymerase II cis-regulatory region sequence-specific DNA binding
DNA-binding transcription factor activity, RNA polymerase II-specific
DNA binding
double-stranded DNA binding
DNA-binding transcription factor activity
protein binding
protein homodimerization activity
bHLH transcription factor binding
sequence-specific DNA binding
protein heterodimerization activity
protein dimerization activity
positive regulation of transcription from RNA polymerase II promoter by glucose
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription by RNA polymerase II
transcription by RNA polymerase II
lactation
positive regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
lipid homeostasis
RNA polymerase II cis-regulatory region sequence-specific DNA binding
DNA-binding transcription factor activity, RNA polymerase II-specific
DNA binding
double-stranded DNA binding
DNA-binding transcription factor activity
protein binding
protein homodimerization activity
bHLH transcription factor binding
sequence-specific DNA binding
protein heterodimerization activity
protein dimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034252 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034253 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034257 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034248 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034259 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034246 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034247 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034254 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034256 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034250 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034251 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034249 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017967 | U01663.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017967 | BC082995.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017967 | U01662.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017967 | AK158906.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017967 | AK146871.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P647919 | U01663.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P647919 | NM_011680.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P647919 | U01663.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P647919 | NM_011680.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 182979 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 934727 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 575884 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 763094 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 493254 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 69419 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 157358 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P120079 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104889 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017967 | U01663.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017967 | BC082995.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017967 | U01662.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017967 | AK158906.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017967 | AK146871.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P647919 | U01663.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P647919 | NM_011680.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P647919 | U01663.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P647919 | NM_011680.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P647919 | U01663.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P647919 | NM_011680.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P647919 | U01663.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P647919 | NM_011680.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P285844 | U01663.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P285844 | BC082995.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P285844 | U01662.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P285844 | AK158906.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P285844 | AB221562.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034211 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034235 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034249 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034196 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034234 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034213 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034245 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034256 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034243 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034239 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034244 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034236 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034199 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034252 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034257 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034238 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034197 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034253 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034250 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034259 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034237 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034240 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034242 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25034247 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P104889 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P120079 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01493873 | - | - |
Whole Mouse Genome G4122A | A_52_P647919 | U01663.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P647919 | NM_011680.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P647919 | U01663.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P647919 | NM_011680.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P285844 | U01663.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P285844 | BC082995.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P285844 | U01662.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P285844 | AK158906.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P285844 | AB221562.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106021 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P104889 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P120079 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06028913 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06028897 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034259 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034250 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034254 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034252 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034249 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034248 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034256 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034253 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034251 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034246 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034247 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25034257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034199 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034230 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034172 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034323 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034325 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034320 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034222 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034213 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034196 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034249 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034236 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034211 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034234 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034245 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034216 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034242 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034141 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034214 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034197 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034219 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034224 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25034229 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06534468 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25034254 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25034250 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25034249 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25034224 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25034239 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05945049 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05945054 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25034232 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25034256 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25034233 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25034199 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25034197 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06009250 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25034204 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06009276 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25034234 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25034228 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25034212 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25034211 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25034257 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25034213 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25034259 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25034223 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25034222 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25034216 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25034230 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25034220 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06009261 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25034219 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25034196 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25034245 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25034226 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10562179 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10562180 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10562174 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10562176 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10562178 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10562177 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10562175 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10562173 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1418241_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1460228_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10562172 | - | - |
Mouse Genome 430 2.0 Array | 1418241_at | - | - |
Mouse Genome 430 2.0 Array | 1460228_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1460228_at | - | - |
Mouse Genome 430 2.0 Array | 1418241_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700024874.mm.1 | TC0700002784.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700024874.mm.1 | NM_011680.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700024874.mm.1 | NM_011680.2 | Refseq |
Mouse Expression Set 430 | 1418241_at | - | - |
Mouse Expression Set 430 | 1460228_at | - | - |
Mouse Expression Set 430 | 1460228_PM_at | - | - |
Mouse Expression Set 430 | 1418241_PM_at | - | - |
Murine Genome U74v2 Set | 103013_at | - | - |
Murine Genome U74v2 Set | 103013_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1460228_at | - | - |
Mouse Genome 430A 2.0 Array | 1418241_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1460228_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1418241_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1460228_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418241_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418241_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1460228_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 532994 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6940039 | - | - |
MouseWG-6 v2 Expression BeadChip | 5570348 | - | - |
MouseWG-6 v2 Expression BeadChip | 3870368 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3870368 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6940039 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5570348 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003870368 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005570348 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006940039 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2513041 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2519634 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1245973 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2519634 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1245973 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2513041 | - | - |
mouse-6 v1.1 (Illumina) | scl0022282.1_25-S | - | - |
mouse-6 v1.1 (Illumina) | scl022282.1_12-S | - | - |
mouse-6 v1.1 (Illumina) | scl022282.6_54-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3870368 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6940039 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5570348 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl022282.6_54-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0022282.1_25-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl022282.1_12-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5570348 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3870368 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6940039 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2519634 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2482756 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2513041 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2482756 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2513041 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2519634 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl022282.6_54-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0022282.1_25-S | - | - |
