Assay Details
Gene Symbol: Rtkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC116341.2 | - | 1446 | 40098130 | 126 | |
BC116342.2 | - | 1446 | 40098131 | 126 | ||
BC158093.1 | - | 11 - 12 | 1449 | 9007380 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
