Assay Details
Gene Symbol: Cftr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021050.2 | NP_066388.1 | 2 - 3 | 303 | 94 | |
GenBank mRNA | AK033621.1 | - | 2 - 3 | 221 | 94 | |
M60493.1 | - | 2 - 3 | 304 | 94 | ||
M69298.1 | - | 2 - 3 | 302 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
