Assay Details
Gene Symbol: Met | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008591.2 | NP_032617.2 | 5 - 6 | 1880 | 113 | |
GenBank mRNA | AK148258.1 | - | 5 - 6 | 1733 | 113 | |
BC117969.1 | - | 3 - 4 | 1524 | 113 | ||
BC117970.1 | - | 3 - 4 | 1520 | 113 | ||
Y00671.1 | - | 3 - 4 | 1525 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
