Assay Details
Gene Symbol: Aoah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012054.4 | NP_036184.1 | 10 - 11 | 1030 | 112 | |
GenBank mRNA | AK084452.1 | - | 10 - 11 | 1025 | 112 | |
AK085343.1 | - | 10 - 11 | 1002 | 112 | ||
AK133301.1 | - | 10 - 11 | 971 | 112 | ||
AK170293.1 | - | 9 - 10 | 566 | 112 | ||
AK171572.1 | - | 10 - 11 | 849 | 112 | ||
BC150767.1 | - | 10 - 11 | 824 | 9087678 | 112 | |
CT010242.1 | - | 10 - 11 | 749 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
