Assay Details
Gene Symbol: Map3k2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011946.3 | NP_036076.2 | 11 - 12 | 1367 | 72 | |
GenBank mRNA | U43186.1 | - | 11 - 12 | 1265 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
