Assay Details
Gene Symbol: Moxd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139296.2 | NP_647457.2 | 12 - 13 | 1616 | 97 | |
GenBank mRNA | AB065134.1 | - | 12 - 13 | 1616 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P201308 | NM_139296.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P201308 | AB065134.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P201308 | NM_139296.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P201308 | AB065134.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 209836 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 880315 | AK005842.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 880315 | AY485267.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 880315 | BC117510.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 880315 | BC117526.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 880315 | NM_027951.1 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 645371 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 204309 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 339249 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 447741 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 472462 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 153006 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156945 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P201308 | NM_139296.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P201308 | AB065134.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P201308 | NM_139296.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P201308 | AB065134.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P201308 | NM_139296.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P201308 | AB065134.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P201308 | NM_139296.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P201308 | AB065134.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391451 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391423 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391420 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391406 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391411 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391427 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391446 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391447 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391431 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391432 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391403 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391453 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391439 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391412 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391442 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391408 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391437 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391404 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391428 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391410 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391400 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391452 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391433 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391425 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391430 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391426 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391402 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391424 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391448 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391414 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391421 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391417 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391444 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391418 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391416 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391436 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24391405 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P156945 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01312122 | - | - |
Whole Mouse Genome G4122A | A_51_P201308 | NM_139296.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P201308 | AB065134.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P201308 | NM_139296.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P201308 | AB065134.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139339 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135596 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P156945 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05671161 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05671164 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05671163 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05671149 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05671156 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05671148 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05671139 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05671153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391336 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391325 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391467 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391426 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391353 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391388 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391366 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391318 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391451 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391372 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391375 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391378 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391465 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391420 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391432 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391363 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391423 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391425 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391485 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391448 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391418 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391356 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391446 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391341 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391416 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391332 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391402 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391339 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391338 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391368 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391317 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391346 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391386 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391439 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391403 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391383 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391394 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391362 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391326 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391470 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391431 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391457 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391364 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391436 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391487 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391444 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391414 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391460 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391478 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391302 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391453 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391347 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24391306 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391414 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391410 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391444 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391383 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05671161 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391402 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05007002 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391426 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391376 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391364 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391395 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391437 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391403 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05357675 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391370 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391448 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391454 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391446 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05065811 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05686117 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391400 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391443 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391368 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391436 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05065784 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391424 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391431 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391360 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391404 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05065787 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05671166 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391393 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391466 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391438 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05357674 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391362 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391465 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391386 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391378 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391379 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391366 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05671148 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391467 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391459 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391451 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391439 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391421 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391388 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391381 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391417 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391399 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391397 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391406 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391427 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391375 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391423 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391425 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391363 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391396 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391412 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391461 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05686119 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391428 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391462 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391432 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05633753 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24391394 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10544299 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544287 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544298 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544291 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544289 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544297 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544290 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544292 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544288 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544294 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544295 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544293 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544296 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1426077_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10544286 | - | - |
Mouse Genome 430 2.0 Array | 1426077_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1426077_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 194357_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 194357_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600016921.mm.1 | TC0600002144.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600016921.mm.1 | NM_139296.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600016921.mm.1 | NM_139296.2 | Refseq |
Mouse Expression Set 430 | 1426077_at | - | - |
Mouse Expression Set 430 | 1426077_PM_at | - | - |
Murine Genome U74v2 Set | 101337_at | - | - |
Murine Genome U74v2 Set | 101337_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1426077_at | - | - |
Mouse Genome 430A 2.0 Array | 1426077_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1426077_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1426077_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 3710064 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003710064 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2771252 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2771252 | - | - |
mouse-6 v1.1 (Illumina) | scl29092.13.1_16-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3710064 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl29092.13.1_16-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3710064 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2862344 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2862344 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2771252 | - | - |
