Assay Details
Gene Symbol: Ace2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130513.1 | NP_001123985.1 | 13 - 14 | 1992 | 70 | |
NM_027286.4 | NP_081562.2 | 12 - 13 | 1844 | 70 | ||
GenBank mRNA | AB053181.1 | - | 12 - 13 | 1848 | 70 | |
AK008530.1 | - | 1 - 2 | 42 | 70 | ||
AK135357.1 | - | 7 - 8 | 1026 | 70 | ||
AK141604.1 | - | 12 - 13 | 1693 | 70 | ||
BC026801.1 | - | 12 - 13 | 1837 | 4236529 | 70 | |
EF408739.1 | - | 13 - 14 | 1953 | 70 | ||
EF408740.1 | - | 13 - 14 | 1953 | 70 | ||
GQ262785.1 | - | 12 - 13 | 1662 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
