Assay Details
Gene Symbol: Ezh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146689.1 | NP_001140161.1 | 11 - 12 | 1521 | 71 | |
NM_007971.2 | NP_031997.2 | 11 - 12 | 1533 | 71 | ||
GenBank mRNA | AK157853.1 | - | 11 - 12 | 1534 | 71 | |
AK220174.1 | - | 11 - 12 | 1488 | 71 | ||
BC003772.1 | - | 11 - 12 | 1442 | 3492110 | 71 | |
BC016391.1 | - | 11 - 12 | 1455 | 3586689 | 71 | |
BC079538.1 | - | 11 - 12 | 1472 | 6817366 | 71 | |
U52951.1 | - | 11 - 12 | 1523 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
