Assay Details
Gene Symbol: Copb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033370.3 | NP_203534.1 | 15 - 16 | 2098 | 73 | |
GenBank mRNA | AF231925.1 | - | 14 - 15 | 2037 | 73 | |
AK135168.1 | - | 15 - 16 | 2130 | 73 | ||
AK149828.1 | - | 15 - 16 | 2116 | 73 | ||
AK172214.1 | - | 15 - 16 | 2100 | 73 | ||
BC030837.1 | - | 15 - 16 | 2098 | 4022216 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
