Assay Details
Gene Symbol: Stat6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009284.2 | NP_033310.2 | 5 - 6 | 767 | 78 | |
GenBank mRNA | AF481808.1 | - | 4 - 5 | 482 | 78 | |
AF481809.1 | - | 4 - 5 | 482 | 78 | ||
AK149563.1 | - | 5 - 6 | 727 | 78 | ||
AK163053.1 | - | 5 - 6 | 768 | 78 | ||
BC127054.1 | - | 5 - 6 | 557 | 40109712 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
