Assay Details
Gene Symbol: Epha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023580.4 | NP_076069.2 | 4 - 5 | 893 | 121 | |
GenBank mRNA | AF131197.1 | - | 4 - 5 | 835 | 121 | |
AK028478.1 | - | 4 - 5 | 894 | 121 | ||
BC071215.1 | - | 4 - 5 | 857 | 4196138 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
