Assay Details
Gene Symbol: Cand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027994.1 | NP_082270.1 | 11 - 12 | 3375 | 73 | |
GenBank mRNA | AK078117.1 | - | 2 - 3 | 602 | 73 | |
AK129225.1 | - | 11 - 12 | 3336 | 73 | ||
BC057457.1 | - | 4 - 5 | 2018 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
