Assay Details
Gene Symbol: Cand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027994.1 | NP_082270.1 | 2 - 3 | 559 | 112 | |
GenBank mRNA | AK129225.1 | - | 2 - 3 | 520 | 112 | |
AK144869.1 | - | 2 - 3 | 551 | 112 | ||
AK158080.1 | - | 2 - 3 | 555 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
