Assay Details
Gene Symbol: Cand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027994.1 | NP_082270.1 | 4 - 5 | 841 | 77 | |
GenBank mRNA | AK129225.1 | - | 4 - 5 | 802 | 77 | |
AK144869.1 | - | 4 - 5 | 833 | 77 | ||
AK158080.1 | - | 4 - 5 | 837 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
