Assay Details
Gene Symbol: Cand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027994.1 | NP_082270.1 | 5 - 6 | 1089 | 155 | |
GenBank mRNA | AK129225.1 | - | 5 - 6 | 1050 | 155 | |
AK144869.1 | - | 5 - 6 | 1081 | 155 | ||
AK158080.1 | - | 5 - 6 | 1085 | 155 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
