Assay Details
Gene Symbol: Eps8l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133191.2 | NP_573454.2 | 14 - 15 | 1446 | 72 | |
GenBank mRNA | BC005492.1 | - | 14 - 15 | 1446 | 3487337 | 72 |
BC009098.1 | - | 13 - 14 | 1730 | 3482190 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
