Assay Details
Gene Symbol: Mgam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171003.1 | NP_001164474.1 | 14 - 15 | 1752 | 99 | |
GenBank mRNA | EU937529.1 | - | 14 - 15 | 1752 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P485850 | AK033548.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P485850 | BC025509.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P485850 | EU937529.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P485850 | NM_001171003.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P485850 | AK031332.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P485850 | AK033548.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P485850 | BC025509.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P485850 | EU937529.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P485850 | NM_001171003.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P485850 | AK031332.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976117 | EU937529.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1976117 | NM_001171003.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 4701 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 798435 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 98731 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 695894 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 830552 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 301556 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 863619 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 727129 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 292819 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 498772 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 757661 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 215494 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 674210 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 530860 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 787656 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593493 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 495277 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 106228 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 907020 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 34346 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 446886 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 730982 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 876266 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 332681 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 387582 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 195986 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 359479 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 848224 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 103568 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 523816 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 940200 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 828892 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 55079 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 706183 | AB028272.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 706183 | AK146657.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 706183 | AK158530.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 706183 | AK167718.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 706183 | AK168220.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 706183 | AK168291.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 706183 | AK185089.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 706183 | AK186924.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 706183 | BC012962.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 706183 | NM_018808.1 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 397443 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654321 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 180101 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 5654 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 132426 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 47447 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 134007 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 140752 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162035 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976117 | EU937529.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1976117 | NM_001171003.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P485850 | AK033548.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P485850 | BC025509.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P485850 | EU937529.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P485850 | NM_001171003.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P485850 | AK031332.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P485850 | AK033548.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P485850 | BC025509.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P485850 | EU937529.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P485850 | NM_001171003.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P485850 | AK031332.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162035 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05874542 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05874603 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05874574 | - | - |
Whole Mouse Genome G4122A | A_52_P485850 | AK033548.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P485850 | BC025509.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P485850 | EU937529.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P485850 | NM_001171003.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P485850 | AK031332.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P485850 | AK033548.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P485850 | BC025509.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P485850 | EU937529.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P485850 | NM_001171003.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P485850 | AK031332.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P291260 | AK033548.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P291260 | BC025509.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P291260 | EU937529.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P291260 | NM_001171003.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162035 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05699422 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24390385 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05699417 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05699429 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24390432 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24390375 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24390395 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05244225 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24390444 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05807447 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24390382 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24390425 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05794752 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24390418 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24390403 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24390424 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24390434 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05244226 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24390408 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05244229 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24390445 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24390405 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24390440 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24390411 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05085402 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24390428 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24390368 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24390429 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05073317 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24390448 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05699416 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24390371 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24390377 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24390370 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05699421 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24390410 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05794750 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05699425 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05807457 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05807443 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24390415 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24390430 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05807441 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05807442 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24390433 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10537516 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10537526 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10537527 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10537513 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10537540 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10537517 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10537542 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10537510 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10537520 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10537541 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10537511 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10537524 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10537523 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10537515 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10537544 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10537519 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10537529 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10537518 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10537543 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10537521 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10537514 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10537512 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10537539 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10537525 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10537530 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10537528 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10537522 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1456777_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10537509 | - | - |
Mouse Genome 430 2.0 Array | 1456777_at | - | - |
Mouse Genome 430 2.0 Array | 1456777_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600003223.mm.1 | TC0600000416.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600003223.mm.1 | NM_001171003.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600003223.mm.1 | NM_001171003.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600003224.mm.1 | TC0600000416.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600003224.mm.1 | NM_001171003.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600003224.mm.1 | NM_001171003.1 | Refseq |
Mouse Expression Set 430 | 1456777_PM_at | - | - |
Mouse Expression Set 430 | 1456777_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1456777_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1456777_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 427438 | - | - |
ABI Mouse Genome Survey Microarray | 717518 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1215251 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1215251 | - | - |
