Assay Details
Gene Symbol: Gm35147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK035726.1 | - | 4 - 5 | 629 | 81 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 39102 |
Gm39102 | -38.3kb | Anti-sense | Chr. 7 - 132420030 - 132423713 on NCBI Build GRCm38 | |
Downstream | ornithine aminotransferase | Oat | AI194874 | 93.8kb | Sense | Chr. 7 - 132557475 - 132576398 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
