Assay Details
Gene Symbol: Copg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017478.3 | NP_059506.1 | 496 | 69 | ||
GenBank mRNA | AF205065.1 | - | 399 | 69 | ||
AJ251067.1 | - | 468 | 69 | |||
AK079599.1 | - | 6 - 7 | 412 | 69 | ||
AK088492.1 | - | 480 | 69 | |||
AK147943.1 | - | 442 | 69 | |||
AK149462.1 | - | 6 - 7 | 433 | 69 | ||
AK163668.1 | - | 6 - 7 | 467 | 69 | ||
BC049178.1 | - | 462 | 6309945 | 69 | ||
BC145898.1 | - | 439 | 40131201 | 69 | ||
BC145900.1 | - | 439 | 40131203 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
