Assay Details
Gene Symbol: Slc26a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289787.1 | NP_001276716.1 | 11 - 12 | 1451 | 98 | |
NM_001289788.1 | NP_001276717.1 | 11 - 12 | 1399 | 98 | ||
NM_030727.5 | NP_109652.3 | 13 - 14 | 1637 | 98 | ||
GenBank mRNA | AF529192.1 | - | 13 - 14 | 1637 | 98 | |
BC108986.1 | - | 11 - 12 | 1391 | 40047724 | 98 | |
BC108987.1 | - | 11 - 12 | 1443 | 40047725 | 98 | |
CJ076475.1 | - | 1 - 2 | 205 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
