Assay Details
Gene Symbol: Phldb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153537.4 | NP_705765.3 | 2 - 3 | 161 | 65 | |
GenBank mRNA | AK214328.1 | - | 1 - 2 | 45 | 65 | |
BC058712.1 | - | 2 - 3 | 157 | 6842596 | 65 | |
BY230078.1 | - | 2 - 3 | 162 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
