Assay Details
Gene Symbol: Lpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145952.1 | NP_001139424.1 | 10 - 11 | 1939 | 77 | |
NM_001145954.1 | NP_001139426.1 | 6 - 7 | 1353 | 77 | ||
NM_178665.5 | NP_848780.3 | 11 - 12 | 2146 | 77 | ||
GenBank mRNA | AK029567.1 | - | 6 - 7 | 1354 | 77 | |
AK040643.1 | - | 9 - 10 | 1700 | 77 | ||
AK045288.1 | - | 11 - 12 | 2007 | 77 | ||
AK051345.1 | - | 11 - 12 | 2018 | 77 | ||
AK054550.1 | - | 10 - 11 | 1942 | 77 | ||
AK076989.1 | - | 6 - 7 | 1344 | 77 | ||
AK136187.1 | - | 1 - 2 | 47 | 77 | ||
AK168431.1 | - | 11 - 12 | 2053 | 77 | ||
AK169780.1 | - | 11 - 12 | 1958 | 77 | ||
BC085321.1 | - | 10 - 11 | 1885 | 30743832 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
