Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600002440.mm.1 |
TC1600000266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600002440.mm.1 |
NM_029436.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600002440.mm.1 |
NM_029436.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600002442.mm.1 |
TC1600000266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600002442.mm.1 |
NM_029436.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600002442.mm.1 |
NM_029436.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600002446.mm.1 |
TC1600000266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600002446.mm.1 |
NM_029436.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600002446.mm.1 |
NM_029436.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600002444.mm.1 |
TC1600000266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600002444.mm.1 |
NM_029436.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600002444.mm.1 |
NM_029436.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600002445.mm.1 |
TC1600000266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600002445.mm.1 |
NM_029436.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600002445.mm.1 |
NM_029436.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600002449.mm.1 |
TC1600000266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600002449.mm.1 |
NM_029436.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600002449.mm.1 |
NM_029436.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600002443.mm.1 |
TC1600000266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600002443.mm.1 |
NM_029436.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600002443.mm.1 |
NM_029436.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600002447.mm.1 |
TC1600000266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600002447.mm.1 |
NM_029436.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600002447.mm.1 |
NM_029436.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600002439.mm.1 |
TC1600000266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600002439.mm.1 |
NM_029436.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600002439.mm.1 |
NM_029436.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600002438.mm.1 |
TC1600000266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600002438.mm.1 |
NM_029436.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600002438.mm.1 |
NM_029436.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600002448.mm.1 |
TC1600000266.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600002448.mm.1 |
NM_029436.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600002448.mm.1 |
NM_029436.3 |
Refseq |