Assay Details
Gene Symbol: Map2k5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011840.2 | NP_035970.1 | 2 - 3 | 772 | 105 | |
GenBank mRNA | AB019374.1 | - | 2 - 3 | 773 | 105 | |
AK020716.1 | - | 2 - 3 | 800 | 105 | ||
AK159261.1 | - | 2 - 3 | 823 | 105 | ||
AK166137.1 | - | 2 - 3 | 823 | 105 | ||
BC013697.1 | - | 2 - 3 | 763 | 105 | ||
BC028260.1 | - | 2 - 3 | 652 | 5067818 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
