Assay Details
Gene Symbol: Sin3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110350.1 | NP_001103820.1 | 13 - 14 | 2373 | 67 | |
NM_001110351.1 | NP_001103821.1 | 14 - 15 | 2250 | 67 | ||
NM_011378.2 | NP_035508.2 | 14 - 15 | 2481 | 67 | ||
GenBank mRNA | AK136347.1 | - | 8 - 9 | 1175 | 67 | |
AK220292.1 | - | 13 - 14 | 2350 | 67 | ||
BC052716.1 | - | 13 - 14 | 2260 | 6837144 | 67 | |
BC078454.1 | - | 13 - 14 | 2234 | 67 | ||
L36831.1 | - | 13 - 14 | 2164 | 67 | ||
L38620.2 | - | 12 - 13 | 2140 | 67 | ||
L38621.2 | - | 12 - 13 | 2140 | 67 | ||
U22394.1 | - | 14 - 15 | 2489 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
