Assay Details
Gene Symbol: Sin3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110350.1 | NP_001103820.1 | 15 - 16 | 3141 | 90 | |
NM_001110351.1 | NP_001103821.1 | 16 - 17 | 3018 | 90 | ||
NM_011378.2 | NP_035508.2 | 16 - 17 | 3249 | 90 | ||
GenBank mRNA | AK136347.1 | - | 10 - 11 | 1943 | 90 | |
AK136972.1 | - | 1 - 2 | 374 | 90 | ||
AK220292.1 | - | 15 - 16 | 3118 | 90 | ||
BC052716.1 | - | 15 - 16 | 3028 | 6837144 | 90 | |
BC053385.1 | - | 15 - 16 | 3011 | 90 | ||
BC078454.1 | - | 15 - 16 | 3002 | 90 | ||
L36831.1 | - | 15 - 16 | 2932 | 90 | ||
L38620.2 | - | 14 - 15 | 2908 | 90 | ||
L38621.2 | - | 14 - 15 | 2908 | 90 | ||
U22394.1 | - | 16 - 17 | 3257 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
