Assay Details
Gene Symbol: Sin3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110350.1 | NP_001103820.1 | 6 - 7 | 1292 | 79 | |
NM_001110351.1 | NP_001103821.1 | 7 - 8 | 1169 | 79 | ||
NM_011378.2 | NP_035508.2 | 7 - 8 | 1400 | 79 | ||
GenBank mRNA | AK032209.1 | - | 6 - 7 | 1160 | 79 | |
AK136347.1 | - | 1 - 2 | 96 | 79 | ||
AK220292.1 | - | 6 - 7 | 1269 | 79 | ||
BC052716.1 | - | 6 - 7 | 1179 | 6837144 | 79 | |
BC053385.1 | - | 6 - 7 | 1162 | 79 | ||
BC078454.1 | - | 6 - 7 | 1153 | 79 | ||
L36831.1 | - | 6 - 7 | 1083 | 79 | ||
L38620.2 | - | 5 - 6 | 1059 | 79 | ||
L38621.2 | - | 5 - 6 | 1059 | 79 | ||
U22394.1 | - | 7 - 8 | 1408 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
