Assay Details
Gene Symbol: Cic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB093224.1 | - | 1 - 2 | 247 | 72 | |
AF363690.1 | - | 6 - 7 | 977 | 72 | ||
AK147276.1 | - | 6 - 7 | 1610 | 72 | ||
AK147535.1 | - | 7 - 8 | 3728 | 72 | ||
BC058665.1 | - | 3 - 4 | 495 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
