Assay Details
Gene Symbol: Fam13c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143776.1 | NP_001137248.1 | 10 - 11 | 1391 | 80 | |
NM_001143777.1 | NP_001137249.1 | 11 - 12 | 1495 | 80 | ||
NM_024244.4 | NP_077206.3 | 10 - 11 | 1391 | 80 | ||
GenBank mRNA | AK004797.1 | - | 10 - 11 | 1375 | 80 | |
AK032053.1 | - | 12 - 13 | 1230 | 80 | ||
AK041147.1 | - | 10 - 11 | 1392 | 80 | ||
AK044049.1 | - | 11 - 12 | 1418 | 80 | ||
AK220421.1 | - | 10 - 11 | 2933 | 80 | ||
BC129839.1 | - | 10 - 11 | 1347 | 40060810 | 80 | |
BC129840.1 | - | 10 - 11 | 1347 | 40060811 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
