Assay Details
Gene Symbol: Ccar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026201.3 | NP_080477.1 | 19 - 20 | 2843 | 80 | |
GenBank mRNA | AF465615.1 | - | 19 - 20 | 2734 | 80 | |
AK077151.1 | - | 3 - 4 | 544 | 80 | ||
AK078424.1 | - | 5 - 6 | 820 | 80 | ||
BC079652.1 | - | 19 - 20 | 2817 | 6844310 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
