Assay Details
Gene Symbol: Ctnna3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164376.1 | NP_001157848.1 | 7 - 8 | 1212 | 80 | |
NM_001164519.1 | NP_001157991.1 | 2 - 3 | 371 | 80 | ||
NM_177612.3 | NP_808280.2 | 7 - 8 | 1140 | 80 | ||
GenBank mRNA | AF344871.1 | - | 7 - 8 | 1212 | 80 | |
AK030166.1 | - | 7 - 8 | 1141 | 80 | ||
AK142745.1 | - | 7 - 8 | 1213 | 80 | ||
BC112409.1 | - | 7 - 8 | 1114 | 40060582 | 80 | |
DQ380429.1 | - | 7 - 8 | 1140 | 80 | ||
DQ380430.1 | - | 2 - 3 | 370 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
