Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900020314.mm.1 |
TC0900002421.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900020314.mm.1 |
NM_001172068.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020314.mm.1 |
NM_001172068.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020314.mm.1 |
NM_001172071.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020314.mm.1 |
NM_001172071.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020311.mm.1 |
TC0900002421.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900020311.mm.1 |
NM_001172068.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020311.mm.1 |
NM_001172068.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020311.mm.1 |
NM_001172069.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020311.mm.1 |
NM_001172069.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020311.mm.1 |
NM_001172070.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020311.mm.1 |
NM_001172070.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020311.mm.1 |
NM_001172071.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020311.mm.1 |
NM_001172071.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020311.mm.1 |
NM_027838.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020311.mm.1 |
NM_027838.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020315.mm.1 |
TC0900002421.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900020315.mm.1 |
NM_001172068.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020315.mm.1 |
NM_001172068.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020315.mm.1 |
NM_001172069.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020315.mm.1 |
NM_001172069.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020315.mm.1 |
NM_001172070.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020315.mm.1 |
NM_001172070.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020315.mm.1 |
NM_001172071.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020315.mm.1 |
NM_001172071.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020315.mm.1 |
NM_027838.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020315.mm.1 |
NM_027838.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020312.mm.1 |
TC0900002421.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900020312.mm.1 |
NM_001172068.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020312.mm.1 |
NM_001172068.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020312.mm.1 |
NM_001172069.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020312.mm.1 |
NM_001172069.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020312.mm.1 |
NM_001172070.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020312.mm.1 |
NM_001172070.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020316.mm.1 |
TC0900002421.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900020316.mm.1 |
NM_001172068.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020316.mm.1 |
NM_001172068.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020316.mm.1 |
NM_001172069.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020316.mm.1 |
NM_001172069.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020316.mm.1 |
NM_001172070.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020316.mm.1 |
NM_001172070.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020316.mm.1 |
NM_001172071.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020316.mm.1 |
NM_001172071.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020316.mm.1 |
NM_027838.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020316.mm.1 |
NM_027838.3 |
Refseq |