Assay Details
Gene Symbol: Abcf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013852.2 | NP_038880.1 | 11 - 12 | 1120 | 69 | |
GenBank mRNA | AK030626.1 | - | 11 - 12 | 1085 | 69 | |
AK177336.1 | - | 3 - 4 | 116 | 69 | ||
AK179781.1 | - | 5 - 6 | 350 | 69 | ||
BC032923.1 | - | 11 - 12 | 1061 | 1430389 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
