Assay Details
Gene Symbol: Robo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175549.4 | NP_780758.3 | 17 - 18 | 2677 | 89 | |
GenBank mRNA | AK085461.1 | - | 18 - 19 | 3313 | 89 | |
AK129396.1 | - | 4 - 5 | 656 | 89 | ||
BC055333.1 | - | 18 - 19 | 3121 | 6409283 | 89 | |
DQ533875.1 | - | 19 - 20 | 2969 | 89 | ||
DQ533876.1 | - | 17 - 18 | 2677 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
