Assay Details
Gene Symbol: Igdcc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290315.1 | NP_001277244.1 | 16 - 17 | 2891 | 65 | |
NM_020043.3 | NP_064427.2 | 16 - 17 | 2888 | 65 | ||
GenBank mRNA | AB052620.1 | - | 16 - 17 | 2888 | 65 | |
AB052621.1 | - | 16 - 17 | 2891 | 65 | ||
AF176694.1 | - | 16 - 17 | 65 | |||
AK122535.1 | - | 2967 | 65 | |||
CB524810.1 | - | 3 - 4 | 350 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
