Assay Details
Gene Symbol: Rtkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081346.1 | NP_001074815.1 | 2 - 3 | 413 | 105 | |
GenBank mRNA | AK030986.1 | - | 2 - 3 | 297 | 105 | |
AK045134.1 | - | 314 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
