Assay Details
Gene Symbol: Ilf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042707.2 | NP_001036172.1 | 11 - 12 | 1507 | 78 | |
NM_001042708.2 | NP_001036173.1 | 12 - 13 | 1546 | 78 | ||
NM_001042709.2 | NP_001036174.1 | 11 - 12 | 1507 | 78 | ||
NM_001277321.1 | NP_001264250.1 | 12 - 13 | 1546 | 78 | ||
NM_001277322.1 | NP_001264251.1 | 13 - 14 | 1474 | 78 | ||
NM_010561.3 | NP_034691.2 | 12 - 13 | 1546 | 78 | ||
GenBank mRNA | AB221609.1 | - | 10 - 11 | 1222 | 78 | |
AF098967.1 | - | 13 - 14 | 1423 | 78 | ||
AF497751.2 | - | 12 - 13 | 1414 | 78 | ||
AF497752.2 | - | 11 - 12 | 1375 | 78 | ||
AK048096.1 | - | 11 - 12 | 1437 | 78 | ||
AK077560.1 | - | 12 - 13 | 1472 | 78 | ||
AK128936.1 | - | 12 - 13 | 1479 | 78 | ||
BC047272.1 | - | 12 - 13 | 1466 | 6440312 | 78 | |
BC060599.1 | - | 9 - 10 | 1112 | 78 | ||
DQ104405.1 | - | 12 - 13 | 1414 | 78 | ||
DQ104406.1 | - | 11 - 12 | 1375 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
